0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Ammonium nitrate
Ammonium nitrate ≥99.999% (metals basis), Puratronic®
Ammonium nitrate ≥99.999% (metals basis), Puratronic®
:
Thermo Fisher Scientific
Ammonium nitrate ≥99.999% (metals basis), Puratronic®
:
Thermo Fisher Scientific
:
6484-52-2
:
:
NH₄NO₃
:
MFCD00011425
:
80.04 g/mol
:
14,534
:
210 °C (dec.)
:
170 °C
:
1.72
:
Ambient
:
229-347-8
:
1942
:
5.1,III
:
GHS03
:
GHS07
Total Metal Impurities:
Max. 0.001%
msds_ALFA010638