ContactUsLinkComponent
Carbon disulphide
  BKEMSO-9056-B005
 :  
undefined
Carbon disulphide
  BKEMSO-9056-B005
 :  
  • Pack type:
    Glass bottle
  • Pk:
    500 ml
  • Seq:
    2725
 :
 :76.14 g/mol
 :13,01821
 :46.2 °C (1013 hPa)
 :–111.6 °C
 :1.2632 g/cm³ (20 °C)
 :–30 °C
 :200-843-6
 :1131
 :3,I
 :GHS02
 :GHS08
 :GHS07
 :Ambient
 :75-15-0
 :CS₂
 :MFCD00011321