Order Entry
Germany
ContactUsLinkComponent
Carbon disulphide ACS, Reag. Ph. Eur. analytical reagent, Supelco®
Flammable
Health
Irritant
Carbon disulphide ACS, Reag. Ph. Eur. analytical reagent, Supelco®
Catalog # 1.02214.1000
Supplier:  Merck
CAS Number:  
undefined
Flammable
Health
Irritant
Carbon disulphide ACS, Reag. Ph. Eur. analytical reagent, Supelco®
Catalog # 1.02214.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    2.1 g/l (20 °C)
  • Melting Point:
    -111.6 °C
  • Molar Mass:
    76.13 g/mol
  • Boiling Point:
    46.5 °C (1013 hPa)
  • Vapor Pressure:
    398 hPa (20 °C)
  • Flash Point:
    -30 °C
  • Explosion Limit:
    1 - 60 %(V)
  • Density:
    1.26 g/cm3 (20 °C)
  • Ignition Point:
    95 °C
  • Evaporation number:
    1.8

Specification Test Results

About this item

 :
 :76.14 g/mol
Merck Index:13,01821
Boiling Pt:46.2 °C (1013 hPa)
Melting Pt:–111.6 °C
Density:1.2632 g/cm³ (20 °C)
Flash Pt:–30 °C
Storage Temperature:Ambient
EINECS:200-843-6
UN:1131
ADR:3 (6.1),I
 :M-Clarity™ quality level = MQ300
 :GHS02
 :GHS08
 :GHS07
CAS Number:75-15-0
Formula:CS₂
MDL Number:MFCD00011321