


Carbon disulphide
Catalog # EHERCA11016000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:76.14 g/mol
Merck Index:13,01821
Boiling Pt:46.2 °C (1013 hPa)
Melting Pt:–111.6 °C
Density:1.2632 g/cm³ (20 °C)
Flash Pt:–30 °C
EINECS:200-843-6
UN:1131
ADR:3,I
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:75-15-0
Formula:CS₂
MDL Number:MFCD00011321