Silica gel, Pore size: 60 Å 0.060-0.200 mm for chromatography
:7631-86-9
:SiO₂
:MFCD00011232
:60.08 g/mol
:Ambient
- Appearance:White Powder
- Particle size:0.060 to 0.200 mm
- Specific gravity:0.46 to 0.55 kg/l
- pH:6.5 to 7.5 (5 % suspension in water)
- Residual water:2 to 4 % (160°C)