Order Entry
Germany
Orders LinkContactUsLinkComponent
Silica gel, granules, TECHNICAL

Silica gel, granules, TECHNICAL

Plastic bottle for solids, 1 kg
Catalog # 27613.294
Supplier:   VWR Chemicals
Silica gel, granules, TECHNICAL
Catalog # 27613.294
Supplier:  VWR Chemicals
CAS Number:  63231-67-4
Silica gel, granules, TECHNICAL
Silica gel, granules, TECHNICAL

Plastic bottle for solids, 1 kg

CAS Number:  63231-67-4
Price
80,80 €
Log in for availability
1 * 1 kg
Product Details & Documents

Moisture absorber, neutral, not deliquescing, not corrosive. It can be regenerated by heating at about 150 °C

  • CAS Number:63231-67-4
  • Formula:SiO₂
  • MDL Number:MFCD00011232
  • Molecular Weight:60.08 g/mol
  • Boiling Pt:2230 °C (1013 hPa)
  • Melting Pt:> 1600 °C
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Loss on drying (140 °C; 4 h):
    Max. 3%
  • Water-absorption capacity (40% RH):
    Min. 20%

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.