


Lead(II) chloride for synthesis, Sigma-Aldrich®
: Merck
:7758-95-4
:
:Cl₂Pb
:MFCD00011157
:278.11 g/mol
:13,05422
:950 °C (1013 hPa)
:496 °C
:5.85 g/cm³ (20 °C)
:Ambient
:2291
:6.1,III
:M-Clarity™ quality level = MQ200
:231-845-5
:GHS08
:GHS07
:GHS09
- Hill Formula:Cl₂Pb
- Chemical formula:PbCl₂
- Molar Mass:278.1 g/mol
- Boiling point:950 °C (1013 hPa)
- Density:5.85 g/cm³ (20 °C)
- Melting Point:500 °C
- Solubility:10 g/l (20 °C)
- LD 50 oral:LD50 Rat 1947 mg/kg
- Storage class:6.1 D Non combustible, acute toxicity cat. 3 / toxic or substances with chronic effects
- WGK:WGK 3 highly water endangering
- R Phrase:R 61-20/22-33-50/53-62
- S Phrase:S 53-45-60-61