Diboron trioxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1303-86-2
:
Formula:B₂O₃
MDL Number:MFCD00011315
:69.62 g/mol
Merck Index:13,01327
Boiling Pt:1860 °C (1013 hPa)
Melting Pt:450 °C
Density:1.81…1.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-125-8
REACH:01-2119486655-24
:GHS08