Diboron trioxide ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1303-86-2
:
Formula:B₂O₃
MDL Number:MFCD00011315
:69.62 g/mol
Merck Index:14,1337
Boiling Pt:1860 °C
Melting Pt:∼450 °C
Density:2.46
Storage Temperature:Ambient
EINECS:215-125-8
REACH:01-2119486655-24
:GHS08