Diboron trioxide 98% -40 mesh
:1303-86-2
:
:B₂O₃
:MFCD00011315
:69.62 g/mol
:13,01327
:1860 °C (760 mmHg)
:450 °C
:2.46 g/cm³
:Ambient
:215-125-8
:01-2119486655-24
:GHS08
- Appearance:White Powder
- Total trace metal impurities:≤0.2 %
- Titration with NaOH:≥97.5 %
- Impurity:≤0.01 % (Total halides impurities)