Palladium 5% on barium sulphate hydrogenation catalyst, reduced
Catalog # STRE46-1989-10G
Supplier: Strem Chemicals, Inc.
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:106.420 g/mol
Storage Temperature:Ambient
EINECS:231-115-6
CAS Number:7440-05-3
Formula:Pd
MDL Number:MFCD00011167