Bis(cyclopentadienyl)ruthenium ≥99% (99.9%-Ru)
Catalog # STRE44-6200-1G
Supplier: Strem Chemicals, Inc.
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:221.18 g/mol
Melting Pt:194…198 °C
EINECS:215-065-2
:GHS07
CAS Number:1287-13-4
Formula:C₁₀Ru
MDL Number:MFCD00001442