Bis(cyclopentadienyl)nickel ≥99%
Catalog # STRE28-1301-5G
Supplier: Strem Chemicals, Inc.
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Melting Pt:173…174 °C
EINECS:215-039-0
ADR:4.1,II
:188.88 g/mol
UN:1325
:GHS02
:GHS06
:GHS08
:GHS07
CAS Number:1271-28-9
Formula:C₁₀H₁₀Ni
MDL Number:MFCD00001441