Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dichlorodiethylsilane
Dichlorodiethylsilane
Dichlorodiethylsilane
Dichlorodiethylsilane
Supplier:
CHEMPUR SILANE
Certificates
Expand image
Dichlorodiethylsilane
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
1719-53-5
:
Formula:
C₄H₁₀Cl₂Si
MDL Number:
MFCD00000493
ADR:
8,II
:
157.11 g/mol
EINECS:
217-005-0
UN:
1767