Bis(cyclopentadienyl)nickel, Sigma-Aldrich®
Catalog # SIALN7524-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
:188.88 g/mol
EINECS:215-039-0
CAS Number:1271-28-9
Formula:C₁₀H₁₀Ni
MDL Number:MFCD00001441