Order Entry
Germany
ContactUsLinkComponent
Platinum, Sigma-Aldrich®
Platinum, Sigma-Aldrich®
Catalog # SIAL373265-550MG
Supplier:  Merck
CAS Number:  
Platinum, Sigma-Aldrich®
Catalog # SIAL373265-550MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    550 mg
  • Seq:
    2770

Specifications

About this item

 :
 :195.08 g/mol
Merck Index:12,07689
Boiling Pt:3827 °C (1013 hPa)
Melting Pt:1769 °C
Density:21.447 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-116-1
CAS Number:7440-06-4
Formula:Pt
MDL Number:MFCD00011179