Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Triethylene glycol monomethyl ether
Triethylene glycol monomethyl ether, Sigma-Aldrich®
Triethylene glycol monomethyl ether, Sigma-Aldrich®
Triethylene glycol monomethyl ether, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Triethylene glycol monomethyl ether, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
112-35-6
:
Formula:
C₇H₁₆O₄
MDL Number:
MFCD00002875
:
164.2 g/mol
:
M-Clarity™ quality level = MQ200
EINECS:
203-962-1