Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dichlorodiethylsilane
Dichlorodiethylsilane, Sigma-Aldrich®
Dichlorodiethylsilane, Sigma-Aldrich®
Dichlorodiethylsilane, Sigma-Aldrich®
SIAL274208-25G
:
Merck
:
Dichlorodiethylsilane, Sigma-Aldrich®
SIAL274208-25G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
8,II
:
M-Clarity™ quality level = MQ100
:
157.11 g/mol
:
217-005-0
:
1767
:
1719-53-5
:
C₄H₁₀Cl₂Si
:
MFCD00000493