
1,4-Butanediol dimethacrylate ≥95% inhibited, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2082-81-7
:
Formula:C₁₂H₁₈O₄
MDL Number:MFCD00008594
:226.27 g/mol
Boiling Pt:132…134 °C (4 mmHg)) (lit.)
Density:1.023 g/mL (25 °C) (lit.)
:M-Clarity™ quality level = MQ100
:Stabilised with 4-methoxyphenol (MEHQ) 200-300 ppm
EINECS:218-218-1
:GHS07
Storage Temperature:Refrigerator