Sodium hydroxymethanesulphinate dihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6035-47-8
:149-44-0
Formula:CH₃NaO₃S·2H₂O
MDL Number:MFCD00150598
:154.12 g/mol
:M-Clarity™ quality level = MQ100
EINECS:205-739-4