Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Anthrone

Glass bottle for solids, 10 g
Catalog # SERA13680.01
Irritant
Marketsource
Supplier:   Serva
Anthrone
Catalog # SERA13680.01
Supplier:  Serva
Marketsource
CAS Number:  90-44-8
Anthrone

Glass bottle for solids, 10 g

CAS Number:  90-44-8
Price
Inquire For Price
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Anthracen-9(10H)-one
  • CAS Number:90-44-8
  • Formula:C₁₄H₁₀O
  • MDL Number:MFCD00001187
  • Molecular Weight:194.23 g/mol
  • EINECS:201-994-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications