
Sodium hydroxymethanesulphinate dihydrate stabilised, SAFC®
Supplier: Merck
Certificates
About this item
CAS Number:6035-47-8
:149-44-0
Formula:CH₃NaO₃S·2H₂O
MDL Number:MFCD00150598
:154.12 g/mol
:Stabilised with sodium carbonate (Na₂CO₃)
EINECS:205-739-4
:GHS08
Storage Temperature:Ambient
Related Information
Certificates
Search by Lot NumberMerck Individual products and tailor-made chemicals for your technical applications
Product Literature
msds_817068