
Sodium hydroxymethanesulphinate dihydrate ≥97.0% stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6035-47-8
:149-44-0
Formula:CH₃NaO₃S·2H₂O
MDL Number:MFCD00150598
:154.12 g/mol
Melting Pt:63 °C
Density:1.8 g/cm3 (20 °C)
:M-Clarity™ quality level = MQ200
:Stabilised with sodium carbonate (Na₂CO₃)
EINECS:205-739-4
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay (iodometric, calc. as dihydrate):≥ 97.0 %
- Iron (Fe):≤ 0.003 %
- Identity (IR):passes test