
Anthrone ≥97.0% for synthesis, Sigma-Aldrich®
8.01461.0010
: Merck
:
- Pack type:Plastic bottle
- Pk:10 g
- Seq:3236
- CAS:90-44-8
- Synonyms:9,10-Dihydro-9-oxoanthracene
- Hill Formula:C₁₄H₁₀O
- Molar Mass:194.23 g/mol
- Melting Point:151 - 156 °C
- Bulk density:450 kg/m³
- Solubility:4.6 mg/l (20 °C)
- Assay (GC, area%):≥ 97,0 Area %
- Identity (IR):conforms
:
:M-Clarity™ quality level = MQ200
:194.23 g/mol
:201-994-0
:GHS07
:Ambient
:90-44-8
:C₁₄H₁₀O
:MFCD00001187