
Ammonia solution 3 mol/l (3 N)
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Storage Temperature:Ambient
EINECS:215-647-6
UN:2672
ADR:8,III
:GHS07
Frequently Bought Together