Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-2-thiazoline hydrochloride
2-Amino-2-thiazoline hydrochloride
2-Amino-2-thiazoline hydrochloride
2-Amino-2-thiazoline hydrochloride
Supplier:
FLUOROCHEM
Certificates
Expand image
2-Amino-2-thiazoline hydrochloride
Supplier:
FLUOROCHEM
About this item
CAS Number:
3882-98-2
:
Formula:
C₃H₆N₂S
MDL Number:
MFCD00012709
:
102.16 g/mol
EINECS:
217-224-1