Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(4-Nitrobenzyl)malononitrile
(4-Nitrobenzyl)malononitrile
(4-Nitrobenzyl)malononitrile
(4-Nitrobenzyl)malononitrile
Supplier:
BIONET RESEARCH
Certificates
Expand image
(4-Nitrobenzyl)malononitrile
Supplier:
BIONET RESEARCH
About this item
CAS Number:
6731-53-9
:
Formula:
C₁₀H₇N₃O₂
MDL Number:
MFCD00231640
:
201.18 g/mol