Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 2-bromonicotinate
Ethyl-2-bromonicotinate 96%
Ethyl-2-bromonicotinate 96%
Irritant
Ethyl-2-bromonicotinate 96%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
Ethyl-2-bromonicotinate 96%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
53087-78-8
:
MFCD04113825
Formula:
C₈H₈BrNO₂
MDL Number:
MFCD11506200
:
230.06 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
msds_ALFAH52335
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation