Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Anthrone 96%

Health
Irritant
Marketsource
Anthrone 96%
Marketsource
Certificates
Anthrone 96%

4 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 90-44-8 Molecular Formula: C14H10O Formula Weight: 194.23 Melting Point: 153-160° Solubility: Soluble in alcohol, benzene, and hot NaOH solutions. Insoluble in water Beilstein Reference: 1910173 Merck Reference: 14,691 MDL No.: MFCD00001187

In organic synthesis, in Colourimetric determination of sugar in body fluids and of animal starch in liver tissue

Synonyms:  Anthracen-9(10H)-one
  • CAS Number:90-44-8
  • Formula:C₁₄H₁₀O
  • MDL Number:MFCD00001187
  • Molecular Weight:194.23 g/mol
  • Merck Index:14,691
  • Melting Pt:153…160 °C
  • Storage Temperature:Ambient
  • EINECS:201-994-0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 85,40 €
    85,40 €
    1 * 25 g
       85,40 €

  • 25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 180,00 €
    180,00 €
    1 * 100 g
       180,00 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 624,00 €
    624,00 €
    1 * 500 g
       624,00 €