Order Entry
Germany
Orders LinkContactUsLinkComponent
Anthrone ACS

Anthrone ACS

Health
Irritant
Anthrone ACS
Certificates
Anthrone ACS

2 Options Available Below

Product Details & Documents

CAS No.: 90-44-8 Molecular Formula: C14H10O Formula Weight: 194.23 Physical Form: Crystalline Sensitivity: Light Sensitive Melting Point: 155-158° Beilstein Reference: 1910173 Merck Reference: 14,691 MDL No.: MFCD00001187

Used for determination of carbohydrates

Synonyms:  Anthracen-9(10H)-one
  • CAS Number:90-44-8
  • Formula:C₁₄H₁₀O
  • MDL Number:MFCD00001187
  • Molecular Weight:194.23 g/mol
  • Merck Index:14,691
  • Melting Pt:155…158 °C
  • Storage Temperature:Ambient
  • EINECS:201-994-0
Product Family Options

Product Information