tert-Butylchlorodimethylsilane 1.0 M in dichloromethane, AcroSeal®
Catalog # 369101000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Colorless Solution
- Infrared spectrum:Conforms
- Titration Argentometric:0.95 to 1.10 M
Specification Test Results
About this item
:
:150.72 g/mol
Flash Pt:58 °C
Storage Temperature:Ambient
EINECS:242-042-4
UN:2920
ADR:8,I
:GHS08
:GHS05
:GHS02
:GHS07
CAS Number:18162-48-6
Formula:C₆H₁₅ClSi
MDL Number:MFCD00000501