Bromoacetaldehyde dimethyl acetal 97% stabilised
:7252-83-7
:
:BrH₂CCH(OCH₃)₂
:MFCD00000213
:169.02 g/mol
:148…150 °C (760 mmHg)
:1.43 g/cm³
:51 °C
:Refrigerator
:230-669-6
:2929
:6.1,II
:GHS06
:GHS02
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4450 to 1.4470 (20°C, 589 nm)
- GC:≥96.0 %
- Stabilizer:0.2 % potassium carbonate