Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cyanoacetamide
Cyanoacetamide 99%
Cyanoacetamide 99%
:
Thermo Fisher Scientific
Cyanoacetamide 99%
:
Thermo Fisher Scientific
:
107-91-5
:
:
C₃H₄N₂O
:
MFCD00008024
:
84.08 g/mol
:
118…122 °C
:
215 °C
:
203-531-8
:
GHS07
:
Ambient
Appearance:
White to cream Crystalline powder
Infrared spectrum:
Conforms
Melting point:
118 to 122 °C
GC:
≥98.5 %
ACRO14865