N-Methylphenethylamine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:589-08-2
:
Formula:C₉H₁₃N
MDL Number:MFCD00008291
:135.21 g/mol
Boiling Pt:203 °C (760 mmHg)
Density:0.93 g/cm³
Flash Pt:73 °C
Storage Temperature:Ambient
EINECS:209-632-3
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5150 to 1.5170 (20°C, 589 nm)
- GC:≥98.5 %