Order Entry
Germany
ContactUsLinkComponent
Cystamine dihydrochloride 97%
Irritant
Cystamine dihydrochloride 97%
Certificates
undefined
Irritant
Cystamine dihydrochloride 97%

Specification Test Results

  • Appearance:
    White to yellow Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥96.0 %
  • Additional info:
    oxalic acid: < 0.05%

Specification Test Results

Related Information