
Sodium hexafluorosilicate
Catalog # HONE01418.9050
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pk:50 kg
- Seq:4418
Specifications
About this item
:
:188.06 g/mol
Merck Index:12,08769
Density:2.68 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:240-934-8
UN:2674
ADR:6.1,III
:GHS06
CAS Number:16893-85-9
Formula:F₆Na₂Si
MDL Number:MFCD00003491