Sodium hydrogen selenite
Catalog # OXOILP0121A
Supplier: THERMO FISHER DIAGNOSTICS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:150.96 g/mol
Storage Temperature:Ambient
EINECS:231-966-3
UN:2630
ADR:6.1,I
:GHS06
:GHS08
:GHS09
CAS Number:7782-82-3
Formula:HNaO₃Se
MDL Number:MFCD00003525