Order Entry
Germany
ContactUsLinkComponent
 
Toxic
Environment
Sodium selenite, TECHNICAL
Catalog # 28074.231
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Environment
Sodium selenite, TECHNICAL
Catalog # 28074.231
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
Merck Index:12,08822
Melting Pt:710 °C
Density:3.1 g/cm³ (20 °C)
ADR:6.1,I
 :172.94 g/mol
EINECS:233-267-9
UN:2630
 :GHS06
 :GHS09
Storage Temperature:Ambient
CAS Number:10102-18-8
Formula:Na₂SeO₃
MDL Number:MFCD00003489