Order Entry
Germany
ContactUsLinkComponent
Sodium selenite
Toxic
Environment
Sodium selenite
Catalog # MOLE28761535-500G
Supplier:  Molekula
CAS Number:  
Toxic
Environment
Sodium selenite
Catalog # MOLE28761535-500G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
Merck Index:12,08822
Melting Pt:710 °C
Density:3.1 g/cm³ (20 °C)
ADR:6.1,I
 :172.94 g/mol
EINECS:233-267-9
UN:2630
 :GHS06
 :GHS09
Storage Temperature:Ambient
CAS Number:10102-18-8
Formula:Na₂SeO₃
MDL Number:MFCD00003489