Sodium hydrogen sulphate, anhydrous ∼95,0% (by titrimetric analysis), Purum, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7681-38-1
:
Formula:HNaO₄S
MDL Number:MFCD00003531
Merck Index:12,08729
Boiling Pt: Dec. (on heating)
Melting Pt:315 °C
Density:2.12 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-665-7
ADR:8,III
:120.06 g/mol
:GHS05
Specification Test Results
- Assay:93.0 - 102.0 %
- Assay of Na₂SO₄:Max. 6 %