Order Entry
Germany
ContactUsLinkComponent
Sodium hydrogen sulphate
Sodium hydrogen sulphate
Catalog # MOLE28194523-1KG
Supplier:  Molekula
CAS Number:  
Sodium hydrogen sulphate
Catalog # MOLE28194523-1KG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
Merck Index:12,08729
Melting Pt:181.85 °C
Density:1.8 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,III
 :120.06 g/mol
EINECS:231-665-7
UN:3260
 :GHS05
CAS Number:7681-38-1
Formula:HNaO₄S
MDL Number:MFCD00003531