Order Entry
Germany
ContactUsLinkComponent
Test tube holder, wood
Test tube holder, wood
Catalog # 241-0415
Supplier:  MARIENFELD
undefined
Test tube holder, wood
Catalog # 241-0415
Supplier:  MARIENFELD

Specifications

  • Length:
    180 mm
  • Pk:
    12
  • Spanning area:
    40 mm

Specifications

About this item

Test tube holder made of wood.

  • With galvanized wire spring
  • For tubes up to 40 mm Ø