Order Entry
Germany
ContactUsLinkComponent
Sodium sulphate ≥99.5%, PESTINORM® for pesticide residue analysis
Sodium sulphate ≥99.5%, PESTINORM® for pesticide residue analysis
Catalog # 28116.293
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium sulphate ≥99.5%, PESTINORM® for pesticide residue analysis
Catalog # 28116.293
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Absence for pesticide residues:
    Passes test
  • pH (20°C; 5 %):
    5.0 - 8.0
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Loss on drying (130°C):
    Max. 0.5 %
  • Loss on ignition (600°C):
    Max. 0.5 %
  • Total N (Nitrogen):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • PO4 (Phosphate):
    Max. 20 ppm
  • As (Arsenic):
    Max. 1 ppm
  • Ca + Mg (as Ca):
    Max. 100 ppm
  • Fe (Iron):
    Max. 2 ppm
  • K (Potassium):
    Max. 100 ppm

Specification Test Results

About this item

CAS Number:7757-82-6
 :
Formula:Na₂SO₄
MDL Number:MFCD00003504
Merck Index:12,08829
Boiling Pt:1429 °C (1013 hPa)
Melting Pt:888 °C
Density:2.7 g/cm³ (20 °C)
 :142.04 g/mol
Storage Temperature:Ambient
EINECS:231-820-9