Order Entry
Germany
ContactUsLinkComponent
Sodium sulphate ≥99%, GPR RECTAPUR®
Sodium sulphate ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Sodium sulphate ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Fe (Iron):
    Max. 20 ppm
  • Sodium chloride:
    Max. 0.25 %

Specification Test Results