Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium sulphate
Sodium sulphate ≥98%, TECHNICAL
Sodium sulphate ≥98%, TECHNICAL
:
VWR Chemicals
Sodium sulphate ≥98%, TECHNICAL
:
VWR Chemicals
:
7757-82-6
:
:
Na₂SO₄
:
MFCD00003504
:
12,08829
:
1429 °C (1013 hPa)
:
888 °C
:
2.7 g/cm³ (20 °C)
:
142.04 g/mol
:
Ambient
:
231-820-9
Assay:
Min. 98 %
1L Plastic bottle for solids DIN 60 UN (ZPACK13251000)
33L Plastic bucket UN Z (ZPACK13247037)
5L Plastic bucket UN Z (ZPACK13247005)
SDS_EN_UK_VWRC28104.pdf
sds_VWRC28104
spec_VWRC28104.292
spec_VWRC28104.361
spec_VWRC28104.460
VWR Chemicals Big Packs and Bulks