Sodium sulphate for synthesis, Sigma-Aldrich®
Catalog # 8.22286.5000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 kg
- Seq:4114
Specifications
Specification Test Results
- Solubility:44.55 g/l (20 °C)
- Melting Point:888 °C
- Molar Mass:142.04 g/mol
- Bulk Density:1400 - 1600 kg/m3
- Density:2.70 g/cm3 (20 °C)
- pH:4 - 7 (200 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
Merck Index:12,08829
Boiling Pt:1429 °C (1013 hPa)
Melting Pt:888 °C
Density:2.7 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:142.04 g/mol
Storage Temperature:Ambient
EINECS:231-820-9
CAS Number:7757-82-6
Formula:Na₂SO₄
MDL Number:MFCD00003504