Order Entry
Germany
Orders LinkContactUsLinkComponent
Sodium bisulphite

Sodium bisulphite

Glass bottle for solids, 500 g
Catalog # WAKO190-02115
Corrosive
Irritant
Marketsource
Supplier:   Wako
Sodium bisulphite
Catalog # WAKO190-02115
Supplier:  Wako
Marketsource
CAS Number:  7631-90-5
Sodium bisulphite

Glass bottle for solids, 500 g

CAS Number:  7631-90-5
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Sodium hydrogen sulphite
  • CAS Number:7631-90-5
  • Formula:HNaO₃S
  • MDL Number:MFCD00167602
  • Molecular Weight:104.06 g/mol
  • Merck Index:12,08731
  • Melting Pt:300 °C
  • Density:2.476 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-548-0
  • UN:2693
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications