Sodium dithionite, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7775-14-6
:
Formula:Na₂S₂O₄
MDL Number:MFCD00011640
:174.11 g/mol
Merck Index:13,08700
Melting Pt:100 °C
Density:2.19 g/cm³ (20 °C)
Flash Pt:101 °C
Storage Temperature:Ambient
EINECS:231-890-0
UN:1384
ADR:4.2,II
:GHS02
:GHS07