Sodium dihydrogen phosphate ≥96%
About this item
Purity: 96% CAS No.: 7558-80-7 Molecular Formula: H2NaO4P Formula Weight: 119.98 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Odour: Odourless Solubility: Freely soluble in water. Insoluble in alcohol Merck Reference: 14,8660 MDL No.: MFCD00003527
- CAS Number:7558-80-7
- Formula:NaH₂PO₄
- MDL Number:MFCD00003527
- Molecular Weight:119.98 g/mol
- Merck Index:14,8660
- Melting Pt:∼650 °C
- Density:0.9996 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-449-2
- REACH:01-2119489796-13
2 Options Available Below
Product Details & Documents
Purity: 96% CAS No.: 7558-80-7 Molecular Formula: H2NaO4P Formula Weight: 119.98 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Odour: Odourless Solubility: Freely soluble in water. Insoluble in alcohol Merck Reference: 14,8660 MDL No.: MFCD00003527
- CAS Number:7558-80-7
- Formula:NaH₂PO₄
- MDL Number:MFCD00003527
- Molecular Weight:119.98 g/mol
- Merck Index:14,8660
- Melting Pt:∼650 °C
- Density:0.9996 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-449-2
- REACH:01-2119489796-13
Dokumente
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



