Disodium hydrogen phosphate, anhydrous 98.0-100.5% (by acidometric titration), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JPE, E339, SAFC®
Supplier: Merck
Certificates
About this item
CAS Number:7558-79-4
:
Formula:Na₂HPO₄
MDL Number:MFCD00003496
:141.96 g/mol
Merck Index:12,08805
Melting Pt:250 °C
Density:1.53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-448-7
REACH:01-2119489797-11
Specification Test Results
- Assay (calculated on dried substance according to Ph Eur):98.0 - 101.0 %
- Assay (calculated on dried substance according to USP):98.0 - 100.5 %
- Assay (acidimetric; dried substance):99.0 - 101.0 %
- Identity:passes test
- Appearance of solution:passes test
- In water insoluble matter (calculated on dried substance):≤ 0.20 %
- Sodium dihydrogen phosphate (NaH₂PO₄):≤ 1.7 %
- pH-value (1 %, water):9.0 - 9.4
- Carbonate:passes test
- Chloride (Cl):≤ 0.01 %
- Fluoride (F):≤ 0.001 %
- Sulfate (SO₄):≤ 0.03 %
- Heavy metals (as Pb):≤ 0.001 %
- As (Arsenic):≤ 0.0001 %
- Cd (Cadmium):≤ 0.0001 %
- Fe (Iron):≤ 0.002 %
- Hg (Mercury):≤ 0.0001 %
- Ni (Nickel)*:≤ 0.0002
- Pb (Lead):≤ 0.0001 %
- Residual solvents (ICH Q3C):excluded by production process
- Substances reducing potassium permanganate:passes test
- Loss on drying (130 °C):≤ 1.0 %
- Particle size (< 0.315 mm):about 99 %
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_106585
SDS
ProductPage_106585 ProductPage_106585